Assay Details
Target Gene Details
Entrez Gene ID: | 80011 |
Gene Name: | family with sequence similarity 192 member A |
Gene Aliases: |
C16orf94, CDA018, CDA10, NIP30 |
Location: |
Chr.16:57152466-57186242 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 7 |
More Information
Set Membership: |
|

Genomic Map