Assay Details
Target Gene Details
Entrez Gene ID: | 9516 |
Gene Name: | lipopolysaccharide induced TNF factor |
Gene Aliases: |
PIG7, SIMPLE, TP53I7 |
Location: |
Chr.16:11547722-11636377 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524676 | Chr.16:11475598 - 11701723 on Build GRCh38 | Loss |
|
| nsv457409 | Chr.16:11449038 - 11728210 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map