Assay Details
Target Gene Details
Entrez Gene ID: | 84516 |
Gene Name: | dynactin subunit 5 |
Gene Aliases: |
- |
Location: |
Chr.16:23641366-23676207 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCTN5 | NM_001199011.1 | NP_001185940.1 | ||
| NM_001199743.1 | NP_001186672.1 | |||
| NM_032486.3 | NP_115875.1 | |||
| NR_037573.1 | ||||
| AK027387.1 | BAB55077.1 | |||
| AK292843.1 | ||||
| AK299631.1 | ||||
| BC004191.2 | AAH04191.1 | |||
| CD388246.1 | ||||
| CR745557.1 | ||||
| DA310703.1 | ||||
| DB087566.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827603 | Chr.16:23643307 - 23644257 on Build GRCh38 | Loss |
|
| esv2714093 | Chr.16:23643283 - 23645183 on Build GRCh38 | Deletion |
|
| nsv819151 | Chr.16:23643337 - 23650806 on Build GRCh38 | Loss |
|
| esv3638233 | Chr.16:23643115 - 23645270 on Build GRCh38 | Loss |
|
| esv2660866 | Chr.16:23643248 - 23645191 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map