Assay Details
Target Gene Details
Entrez Gene ID: | 10200 |
Gene Name: | M-phase phosphoprotein 6 |
Gene Aliases: |
MPP, MPP-6, MPP6 |
Location: |
Chr.16:82148162-82170224 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPHOSPH6 | NM_005792.2 | NP_005783.2 | ||
| XM_011522808.2 | XP_011521110.1 | |||
| XM_017022852.1 | XP_016878341.1 | |||
| AK222667.1 | BAD96387.1 | |||
| AK314163.1 | ||||
| AW175781.1 | ||||
| BC005242.1 | AAH05242.1 | |||
| BC011020.1 | AAH11020.1 | |||
| BC031017.1 | AAH31017.1 | |||
| BU595100.1 | ||||
| X98263.1 | CAA66916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3050n100 | Chr.16:82131709 - 82181783 on Build GRCh38 | Loss |
|
| nsv573371 | Chr.16:82131578 - 83629456 on Build GRCh38 | Loss |
|
| dgv5251n54 | Chr.16:82102961 - 82183017 on Build GRCh38 | Loss |
|
| esv3639355 | Chr.16:82149872 - 82196025 on Build GRCh38 | Loss |
|
| nsv518129 | Chr.16:82154568 - 82194471 on Build GRCh38 | Loss |
|
| nsv833302 | Chr.16:82154746 - 82330258 on Build GRCh38 | Loss |
|
| esv3639354 | Chr.16:82134351 - 82201089 on Build GRCh38 | Gain |
|
| nsv573367 | Chr.16:82106956 - 83007241 on Build GRCh38 | Gain |
|
| nsv1061288 | Chr.16:82146152 - 83636829 on Build GRCh38 | Gain+Loss |
|
| dgv3051n100 | Chr.16:82138730 - 83654472 on Build GRCh38 | Gain |
|
| nsv525467 | Chr.16:82121014 - 82189880 on Build GRCh38 | Loss |
|
| dgv5253n54 | Chr.16:82148499 - 83630977 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74249007] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map