Assay Details
Target Gene Details
Entrez Gene ID: | 10595 |
Gene Name: | endoplasmic reticulum to nucleus signaling 2 |
Gene Aliases: |
IRE1-BETA, IRE1b, IRE2p, hIRE2p |
Location: |
Chr.16:23690304-23713500 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERN2 | NM_001308220.1 | NP_001295149.2 | ||
| NM_033266.3 | NP_150296.4 | |||
| XM_011545708.1 | XP_011544010.1 | |||
| XM_011545709.1 | XP_011544011.1 | |||
| AB047079.1 | BAB21297.1 | |||
| AK131280.1 | BAD18455.1 | |||
| EF560720.1 | ||||
| EF560739.1 |
More Information
Set Membership: |
|

Genomic Map