Assay Details
Target Gene Details
Entrez Gene ID: | 6645 |
Gene Name: | syntrophin beta 2 |
Gene Aliases: |
D16S2531E, EST25263, SNT2B2, SNT3, SNTL |
Location: |
Chr.16:69186133-69309052 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNTB2 | NM_006750.3 | NP_006741.1 | ||
| AF243385.1 | AAK15149.1 | |||
| BC048215.1 | AAH48215.1 | |||
| U40572.1 | AAC50449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509628 | Chr.16:69175927 - 69248185 on Build GRCh38 | Insertion |
|
| esv2714661 | Chr.16:69196190 - 69211384 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map