Assay Details
Target Gene Details
Entrez Gene ID: | 6340 |
Gene Name: | sodium channel epithelial 1 gamma subunit |
Gene Aliases: |
BESC3, ENaCg, ENaCgamma, PHA1, SCNEG |
Location: |
Chr.16:23182719-23216879 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCNN1G | NM_001039.3 | NP_001030.2 | ||
| BC059391.1 | AAH59391.1 | |||
| BC069652.1 | AAH69652.1 | |||
| DQ402524.1 | ||||
| L36592.1 | AAA75460.1 | |||
| X87160.1 | CAA60633.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77684620] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map