Assay Details
Target Gene Details
Entrez Gene ID: | 102724084 |
Gene Name: | uncharacterized LOC102724084 |
Gene Aliases: |
- |
Location: |
Chr.16:80155958-80563135 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724084 | NR_120307.1 | |||
| DA228026.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833298 | Chr.16:80088440 - 80280706 on Build GRCh38 | Gain |
|

Genomic Map