Assay Details
Target Gene Details
Entrez Gene ID: | 27327 |
Gene Name: | trinucleotide repeat containing 6A |
Gene Aliases: |
CAGH26, GW1, GW182, TNRC6 |
Location: |
Chr.16:24610260-24826227 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNRC6A | XM_017023147.1 | XP_016878636.1 | ||
| U80739.1 | AAB91438.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs80263423] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map