Assay Details
Target Gene Details
Entrez Gene ID: | 9093 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member A3 |
Gene Aliases: |
HCA57, TID1, hTID-1 |
Location: |
Chr.16:4425805-4456775 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJA3 | NM_001135110.2 | NP_001128582.1 | ||
| NM_001286516.1 | NP_001273445.1 | |||
| NM_005147.5 | NP_005138.3 | |||
| AB209923.1 | BAD93160.1 | |||
| AF061749.1 | AAC29066.1 | |||
| AF244136.1 | AAF66245.1 | |||
| AF411044.1 | AAL35323.1 | |||
| AK001333.1 | ||||
| AK223105.1 | BAD96825.1 | |||
| AK295391.1 | ||||
| AK314218.1 | ||||
| BC007225.1 | AAH07225.1 | |||
| BC011855.2 | AAH11855.1 | |||
| BC012343.1 | AAH12343.1 | |||
| BC014062.2 | AAH14062.1 | |||
| BC030145.2 | AAH30145.1 | |||
| BC032100.2 | AAH32100.1 | |||
| BX445320.2 | ||||
| DA172592.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160332 | Chr.16:4367643 - 4486554 on Build GRCh38 | Duplication |
|
| nsv833132 | Chr.16:4370092 - 4463448 on Build GRCh38 | Loss |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv571274 | Chr.16:4309420 - 4441189 on Build GRCh38 | Gain |
|
| esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain |
|
| esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78115271] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map