Assay Details
Target Gene Details
Entrez Gene ID: | 123879 |
Gene Name: | defective in cullin neddylation 1 domain containing 3 |
Gene Aliases: |
44M2.4, SCCRO3 |
Location: |
Chr.16:20854925-20900358 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCUN1D3 | NM_173475.3 | NP_775746.1 | ||
| BC040442.1 | AAH40442.1 | |||
| BX648998.1 | ||||
| DA545688.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638159 | Chr.16:20882859 - 20891671 on Build GRCh38 | Loss |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map