Assay Details
Target Gene Details
Entrez Gene ID: | 1327 |
Gene Name: | cytochrome c oxidase subunit 4I1 |
Gene Aliases: |
COX IV-1, COX4, COX4-1, COXIV, COXIV-1 |
Location: |
Chr.16:85799567-85807003 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX4I1 | NM_001318786.1 | NP_001305715.1 | ||
| NM_001318788.1 | NP_001305717.1 | |||
| NM_001318794.1 | NP_001305723.1 | |||
| NM_001318797.1 | NP_001305726.1 | |||
| NM_001318802.1 | NP_001305731.1 | |||
| NM_001861.4 | NP_001852.1 | |||
| AK311847.1 | ||||
| BC008704.1 | AAH08704.1 | |||
| BC021236.2 | AAH21236.1 | |||
| BC047869.1 | AAH47869.1 | |||
| BC062437.1 | AAH62437.1 | |||
| BE615615.1 | ||||
| BE728651.1 | ||||
| BG614492.1 | ||||
| BM835194.1 | ||||
| BP343428.1 | ||||
| BT019825.1 | AAV38628.1 | |||
| CD677397.1 | ||||
| CN406213.1 | ||||
| DA623889.1 | ||||
| DB502762.1 | ||||
| M21575.1 | AAA99312.1 | |||
| M34600.1 | AAA52059.1 | |||
| U90915.1 | AAB51058.1 | |||
| X54802.1 | CAA38573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833316 | Chr.16:85697332 - 85900033 on Build GRCh38 | Loss |
|
| nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map