Assay Details
Target Gene Details
Entrez Gene ID: | 10143 |
Gene Name: | C-type lectin domain family 3 member A |
Gene Aliases: |
CLECSF1 |
Location: |
Chr.16:78022546-78032106 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC3A | NM_005752.4 | NP_005743.4 | ||
| AK310847.1 | ||||
| AK313316.1 | ||||
| AY358376.1 | AAQ88742.1 | |||
| BC104414.2 | AAI04415.1 | |||
| BC104415.2 | AAI04416.1 | |||
| DA998903.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582336 | Chr.16:78011228 - 78081777 on Build GRCh38 | Loss |
|
| dgv3032n100 | Chr.16:77984145 - 78046521 on Build GRCh38 | Loss |
|
| esv3582337 | Chr.16:78011619 - 78043185 on Build GRCh38 | Loss |
|
| nsv1058985 | Chr.16:78026370 - 78064108 on Build GRCh38 | Loss |
|
| dgv5216n54 | Chr.16:77986647 - 78037610 on Build GRCh38 | Loss |
|
| esv3639185 | Chr.16:78008451 - 78045266 on Build GRCh38 | Loss |
|
| dgv3031n100 | Chr.16:77892138 - 78561070 on Build GRCh38 | Gain |
|
| nsv1056016 | Chr.16:77990656 - 78129245 on Build GRCh38 | Loss |
|
| nsv833293 | Chr.16:77932757 - 78134791 on Build GRCh38 | Gain |
|
| esv2760418 | Chr.16:77894064 - 78560274 on Build GRCh38 | Gain+Loss |
|
| esv3639187 | Chr.16:78025112 - 78042023 on Build GRCh38 | Gain |
|
| nsv1056561 | Chr.16:78002905 - 78070733 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117090245,rs75240778] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map