Assay Details
Target Gene Details
Entrez Gene ID: | 1773 |
Gene Name: | deoxyribonuclease I |
Gene Aliases: |
DNL1, DRNI |
Location: |
Chr.16:3611705-3665472 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNASE1 | XM_005255148.3 | XP_005255205.1 | ||
| XM_006720854.3 | XP_006720917.1 | |||
| XM_011522393.2 | XP_011520695.1 | |||
| XM_017022992.1 | XP_016878481.1 | |||
| XM_017022993.1 | XP_016878482.1 | |||
| XM_017022994.1 | XP_016878483.1 | |||
| XM_017022995.1 | XP_016878484.1 | |||
| XM_017022997.1 | XP_016878486.1 | |||
| XM_017023001.1 | XP_016878490.1 | |||
| XM_017023004.1 | XP_016878493.1 | |||
| XM_017023005.1 | XP_016878494.1 | |||
| XM_017023007.1 | XP_016878496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833128 | Chr.16:3525122 - 3690497 on Build GRCh38 | Loss |
|
| esv2668028 | Chr.16:3612227 - 3634269 on Build GRCh38 | Deletion |
|
| nsv518509 | Chr.16:3583656 - 3688960 on Build GRCh38 | Gain |
|
| esv3637670 | Chr.16:3612113 - 3634450 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map