Assay Details
Target Gene Details
Entrez Gene ID: | 54920 |
Gene Name: | dihydrouridine synthase 2 |
Gene Aliases: |
DUS2L, SMM1, URLC8 |
Location: |
Chr.16:68022944-68079323 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUS2 | NM_001271762.1 | NP_001258691.1 | ||
| NM_001271763.1 | NP_001258692.1 | |||
| NM_017803.4 | NP_060273.1 | |||
| AB101210.1 | BAE07219.1 | |||
| AK000406.1 | BAA91143.1 | |||
| AK290578.1 | ||||
| AK296663.1 | ||||
| BC006527.2 | AAH06527.1 | |||
| DC397403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2666619 | Chr.16:68003989 - 68045407 on Build GRCh38 | Deletion |
|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| esv3638897 | Chr.16:68004075 - 68045657 on Build GRCh38 | Loss |
|
| esv3638901 | Chr.16:68024016 - 68030339 on Build GRCh38 | Loss |
|
| esv3638899 | Chr.16:68008886 - 68035332 on Build GRCh38 | Loss |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map