Assay Details
Target Gene Details
Entrez Gene ID: | 7567 |
Gene Name: | zinc finger protein 19 |
Gene Aliases: |
KOX12 |
Location: |
Chr.16:71474073-71489351 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF19 | NM_006961.3 | NP_008892.2 | ||
| AK057330.1 | BAB71430.1 | |||
| AK292260.1 | ||||
| BC047091.1 | AAH47091.1 | |||
| DB079697.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs545402974] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map