Assay Details
Target Gene Details
Entrez Gene ID: | 1258 |
Gene Name: | cyclic nucleotide gated channel beta 1 |
Gene Aliases: |
CNCG2, CNCG3L, CNCG4, CNG4, CNGB1B, GAR1, GARP, GARP2, RCNC2, RCNCb, RCNCbeta, RP45 |
Location: |
Chr.16:57882340-57971116 on Build GRCh38 |
Assay Gene Location: | Within Intron 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNGB1 | NM_001286130.1 | NP_001273059.1 | ||
| NM_001297.4 | NP_001288.3 | |||
| XM_011522870.2 | XP_011521172.1 | |||
| AF042498.1 | AAC04830.1 | |||
| U58837.1 | AAB63387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062660 | Chr.16:57848645 - 57945049 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map