Assay Details
Target Gene Details
Entrez Gene ID: | 6338 |
Gene Name: | sodium channel epithelial 1 beta subunit |
Gene Aliases: |
BESC1, ENaCb, ENaCbeta, SCNEB |
Location: |
Chr.16:23278231-23381299 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCNN1B | NM_000336.2 | NP_000327.2 | ||
| XM_011545913.2 | XP_011544215.1 | |||
| XM_017023525.1 | XP_016879014.1 | |||
| XM_017023526.1 | XP_016879015.1 | |||
| AF260226.1 | AAK49394.1 | |||
| AK313192.1 | ||||
| BC036352.1 | AAH36352.2 | |||
| X87159.1 | CAA60632.1 |
More Information
Set Membership: |
|

Genomic Map