Assay Details
Target Gene Details
Entrez Gene ID: | 29105 |
Gene Name: | cilia and flagella associated protein 20 |
Gene Aliases: |
C16orf80, EVORF, GTL3, fSAP23 |
Location: |
Chr.16:58113588-58129425 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP20 | NM_013242.2 | NP_037374.1 | ||
| NR_136632.1 | ||||
| AF087845.1 | AAP97158.1 | |||
| AF093680.1 | AAD40382.1 | |||
| AK027099.1 | ||||
| AK297819.1 | ||||
| AV751497.1 | ||||
| BC005152.2 | AAH05152.1 | |||
| CN364433.1 | ||||
| HY042096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638720 | Chr.16:58086282 - 58146835 on Build GRCh38 | Gain |
|
| esv3638719 | Chr.16:58084081 - 58175175 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73549156] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map