Assay Details
Target Gene Details
Entrez Gene ID: | 55808 |
Gene Name: | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1 |
Gene Aliases: |
HSY11339, SIAT7A, ST6GalNAcI, STYI |
Location: |
Chr.17:76617769-76643838 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GALNAC1 | XM_011524996.2 | 9 | 1829 | XP_011523298.1 |
| XM_017024842.1 | 10 | 1943 | XP_016880331.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060182 | Chr.17:76597910 - 76660156 on Build GRCh38 | Gain |
|
| nsv518385 | Chr.17:76597821 - 76661078 on Build GRCh38 | Gain |
|
| nsv1058486 | Chr.17:76592348 - 76634542 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map