Assay Details
Target Gene Details
Entrez Gene ID: | 284085 |
Gene Name: | keratin 18 pseudogene 55 |
Gene Aliases: |
- |
Location: |
Chr.17:28275986-28307382 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT18P55 | NR_028334.1 | |||
| AK097823.1 | BAC05178.1 | |||
| BC112026.1 | AAI12027.1 | |||
| BC112028.1 | AAI12029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519330 | Chr.17:28283642 - 28303453 on Build GRCh38 | Gain |
|
| nsv574653 | Chr.17:28227749 - 28501479 on Build GRCh38 | Gain |
|

Genomic Map