Assay Details
Target Gene Details
Entrez Gene ID: | 8811 |
Gene Name: | galanin receptor 2 |
Gene Aliases: |
GAL2-R, GALNR2, GALR-2 |
Location: |
Chr.17:76070822-76077541 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALR2 | XM_011525427.2 | XP_011523729.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2142 | Chr.17:76070185 - 76087802 on Build GRCh38 | Insertion |
|
| nsv833546 | Chr.17:76003077 - 76186972 on Build GRCh38 | Loss |
|
| esv989375 | Chr.17:76072608 - 76073847 on Build GRCh38 | Insertion |
|
| nsv509674 | Chr.17:76052197 - 76106436 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map