Assay Details
Target Gene Details
Entrez Gene ID: | 100133991 |
Gene Name: | MAP3K14 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:45247925-45268630 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K14-AS1 | NR_024434.2 | |||
| NR_110325.1 | ||||
| NR_110326.1 | ||||
| AK311429.1 | ||||
| AL710937.1 | ||||
| DB133288.1 | ||||
| DB294632.1 |
Target Gene Details
Entrez Gene ID: | 124783 |
Gene Name: | spermatogenesis associated 32 |
Gene Aliases: |
AEP2, C17orf46, TEX34, VAD1.2 |
Location: |
Chr.17:45254393-45262112 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA32 | NM_152343.2 | NP_689556.2 | ||
| AF433663.1 | AAP97314.1 | |||
| AK058143.1 | BAB71683.1 | |||
| AK302184.1 | ||||
| BC028124.1 | AAH28124.1 | |||
| BP369808.1 | ||||
| BP370574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953904 | Chr.17:45231034 - 45277033 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72832885] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map