Assay Details
Target Gene Details
Entrez Gene ID: | 100190938 |
Gene Name: | RAMP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:42753914-42761257 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAMP2-AS1 | NR_024461.1 | 3 | 738 | |
| NR_024462.1 | 4 | 617 | ||
| AK057121.1 | 2 | 766 | ||
| BC001607.1 | 3 | 584 | AAH01607.1 | |
| BC018638.2 | 4 | 617 | AAH18638.2 | |
| BC033118.1 | 4 | 617 | AAH33118.1 | |
| BC034924.1 | 3 | 492 | AAH34924.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833450 | Chr.17:42660985 - 42825144 on Build GRCh38 | Loss |
|

Genomic Map