Assay Details
Target Gene Details
Entrez Gene ID: | 535 |
Gene Name: | ATPase H+ transporting V0 subunit a1 |
Gene Aliases: |
ATP6N1, ATP6N1A, Stv1, VPP1, Vph1, a1 |
Location: |
Chr.17:42458844-42522579 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
| nsv575051 | Chr.17:42317580 - 42498404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map