Assay Details
Target Gene Details
Entrez Gene ID: | 100505753 |
Gene Name: | keratin associated protein 16-1 |
Gene Aliases: |
KAP16.1 |
Location: |
Chr.17:41307700-41309253 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP16-1 | NM_001146182.1 | 1 | 652 | NP_001139654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055426 | Chr.17:41266750 - 41322116 on Build GRCh38 | Gain |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|

Genomic Map