Assay Details
Target Gene Details
Entrez Gene ID: | 51747 |
Gene Name: | LUC7 like 3 pre-mRNA splicing factor |
Gene Aliases: |
CRA, CREAP-1, CROP, LUC7A, OA48-18, hLuc7A |
Location: |
Chr.17:50719565-50754269 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUC7L3 | NM_006107.3 | NP_006098.2 | ||
| NM_016424.4 | NP_057508.2 | |||
| XM_005257448.1 | XP_005257505.1 | |||
| XM_005257449.2 | XP_005257506.1 | |||
| XM_005257452.1 | XP_005257509.1 | |||
| XM_006721943.2 | XP_006722006.1 | |||
| AK001925.1 | BAA91981.1 | |||
| AK023672.1 | ||||
| AK225980.1 | ||||
| AK290540.1 | ||||
| AK295983.1 | ||||
| BC002925.2 | AAH02925.1 | |||
| BC047043.1 | AAH47043.1 | |||
| BC056409.1 | AAH56409.1 | |||
| DA231851.1 | ||||
| DQ013876.1 | AAY26238.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv42n68 | Chr.17:50584511 - 50823099 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map