Assay Details
Target Gene Details
Entrez Gene ID: | 22901 |
Gene Name: | arylsulfatase G |
Gene Aliases: |
- |
Location: |
Chr.17:68259089-68452000 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARSG | NM_014960.4 | NP_055775.2 | ||
| XM_005257170.3 | XP_005257227.1 | |||
| XM_005257172.3 | XP_005257229.1 | |||
| XM_006721777.3 | XP_006721840.2 | |||
| XM_006721779.3 | XP_006721842.1 | |||
| XM_011524535.2 | XP_011522837.1 | |||
| XM_011524538.2 | XP_011522840.1 | |||
| XM_011524540.2 | XP_011522842.1 | |||
| XM_011524541.2 | XP_011522843.1 | |||
| XM_011524542.2 | XP_011522844.1 | |||
| XM_011524543.2 | XP_011522845.1 | |||
| XM_011524544.2 | XP_011522846.1 | |||
| XM_011524545.2 | XP_011522847.1 | |||
| XM_017024361.1 | XP_016879850.1 | |||
| XM_017024366.1 | XP_016879855.1 | |||
| AY358380.1 | AAQ88746.1 |
Target Gene Details
Entrez Gene ID: | 9120 |
Gene Name: | solute carrier family 16 member 6 |
Gene Aliases: |
MCT6, MCT7 |
Location: |
Chr.17:68267026-68291264 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A6 | NM_001174166.1 | NP_001167637.1 | ||
| NM_004694.4 | NP_004685.2 | |||
| AK290417.1 | ||||
| AK313121.1 | ||||
| AX750581.1 | CAD99143.1 | |||
| BC064832.1 | AAH64832.1 | |||
| U79745.1 | AAC52014.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76258556] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map