Assay Details
Target Gene Details
Entrez Gene ID: | 146850 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 6 |
Gene Aliases: |
C17orf38, HsT41028, p84 PIKAP, p87(PIKAP), p87PIKAP |
Location: |
Chr.17:8802723-8870003 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3R6 | NM_001010855.3 | NP_001010855.1 | ||
| NM_001290211.1 | NP_001277140.1 | |||
| NR_110865.1 | ||||
| XM_011523677.2 | XP_011521979.1 | |||
| XM_011523678.2 | XP_011521980.1 | |||
| XM_011523680.2 | XP_011521982.1 | |||
| XM_011523681.2 | XP_011521983.1 | |||
| XM_011523683.2 | XP_011521985.1 | |||
| XM_011523686.2 | XP_011521988.1 | |||
| XM_017024228.1 | XP_016879717.1 | |||
| XM_017024229.1 | XP_016879718.1 | |||
| XM_017024230.1 | XP_016879719.1 | |||
| XM_017024231.1 | XP_016879720.1 | |||
| AK091819.1 | ||||
| AY753192.1 | AAV30085.1 | |||
| BC125169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1969 | Chr.17:8780681 - 8825867 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map