Assay Details
Target Gene Details
Entrez Gene ID: | 5608 |
Gene Name: | mitogen-activated protein kinase kinase 6 |
Gene Aliases: |
MAPKK6, MEK6, MKK6, PRKMK6, SAPKK-3, SAPKK3 |
Location: |
Chr.17:69414697-69553854 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP2K6 | NM_002758.3 | NP_002749.2 | ||
| AB451252.1 | ||||
| AB451376.1 | ||||
| AK225719.1 | ||||
| AK290747.1 | ||||
| BC012009.1 | AAH12009.1 | |||
| D87905.1 | BAA13496.1 | |||
| DB111443.1 | ||||
| U39065.1 | AAB03705.1 | |||
| U39656.1 | AAC50388.1 | |||
| U39657.1 | AAC50389.1 | |||
| U49732.1 | AAB05035.1 | |||
| X96757.1 | CAA65532.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2130 | Chr.17:69413467 - 69445713 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map