Assay Details
Target Gene Details
Entrez Gene ID: | 51195 |
Gene Name: | Rap guanine nucleotide exchange factor like 1 |
Gene Aliases: |
Link-GEFII |
Location: |
Chr.17:40177010-40195656 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAPGEFL1 | NM_001303533.1 | NP_001290462.1 | ||
| NM_001303534.1 | NP_001290463.1 | |||
| NM_016339.4 | NP_057423.1 | |||
| XM_006721932.3 | XP_006721995.1 | |||
| XM_006721935.3 | XP_006721998.1 | |||
| XM_006721937.3 | 1 | 212 | XP_006722000.1 | |
| AF117946.1 | AAF22003.1 | |||
| AK304160.1 | ||||
| AK313492.1 | ||||
| DB265550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525796 | Chr.17:40150112 - 40248785 on Build GRCh38 | Gain |
|
| nsv833444 | Chr.17:40086570 - 40281028 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map