Assay Details
Target Gene Details
Entrez Gene ID: | 101927911 |
Gene Name: | uncharacterized LOC101927911 |
Gene Aliases: |
- |
Location: |
Chr.17:2962247-2965895 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927911 | NR_110818.1 | 1 | 1184 |
Target Gene Details
Entrez Gene ID: | 23108 |
Gene Name: | RAP1 GTPase activating protein 2 |
Gene Aliases: |
GARNL4, RAP1GA3 |
Location: |
Chr.17:2755698-3037741 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP1GAP2 | NM_001100398.1 | NP_001093868.1 | ||
| NM_015085.4 | NP_055900.4 | |||
| XM_005256542.2 | XP_005256599.1 | |||
| XM_006721477.3 | XP_006721540.1 | |||
| XM_011523738.1 | XP_011522040.1 | |||
| XM_011523739.1 | XP_011522041.1 | |||
| XM_011523740.2 | XP_011522042.1 | |||
| XM_011523741.2 | XP_011522043.1 | |||
| XM_011523742.2 | XP_011522044.1 | |||
| XM_011523743.2 | XP_011522045.1 | |||
| XM_017024370.1 | XP_016879859.1 | |||
| XM_017024371.1 | XP_016879860.1 | |||
| AJ628446.1 | CAF31652.1 | |||
| AJ628447.1 | CAF31653.1 | |||
| AK124640.1 | BAC85912.1 | |||
| BC140870.1 | ||||
| BC140871.1 | ||||
| CR627427.1 | CAH10514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain |
|
| esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion |
|
| nsv574227 | Chr.17:2924664 - 3036687 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map