Assay Details
Target Gene Details
Entrez Gene ID: | 374786 |
Gene Name: | EF-hand calcium binding domain 5 |
Gene Aliases: |
- |
Location: |
Chr.17:29929856-30108452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB5 | NM_001145053.1 | NP_001138525.1 | ||
| AK299608.1 | ||||
| AK302745.1 | ||||
| AK303060.1 |
More Information
Set Membership: |
|

Genomic Map