Assay Details
Target Gene Details
Entrez Gene ID: | 6341 |
Gene Name: | SCO1 cytochrome c oxidase assembly protein |
Gene Aliases: |
SCOD1 |
Location: |
Chr.17:10679770-10697568 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCO1 | NM_004589.3 | NP_004580.1 | ||
| XM_005256751.3 | XP_005256808.1 | |||
| AF026852.1 | AAD08641.1 | |||
| AF131816.1 | AAD20051.1 | |||
| AF183424.1 | AAG09693.1 | |||
| AK074588.1 | ||||
| AK098826.1 | ||||
| AK315595.1 | ||||
| BC015504.2 | AAH15504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065819 | Chr.17:10622619 - 10784575 on Build GRCh38 | Gain |
|
| nsv1063900 | Chr.17:10662646 - 10729182 on Build GRCh38 | Gain |
|
| nsv833367 | Chr.17:10683841 - 10826633 on Build GRCh38 | Loss |
|
| nsv1058835 | Chr.17:10322963 - 10939602 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map