Assay Details
Target Gene Details
Entrez Gene ID: | 55813 |
Gene Name: | UTP6, small subunit processome component |
Gene Aliases: |
C17orf40, HCA66 |
Location: |
Chr.17:31862379-31901710 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UTP6 | NM_018428.2 | NP_060898.2 | ||
| XM_005258012.2 | XP_005258069.1 | |||
| XM_011524997.2 | XP_011523299.1 | |||
| XM_017024845.1 | XP_016880334.1 | |||
| AF116631.1 | AAF71054.1 | |||
| AF244135.1 | AAF66244.1 | |||
| AF334386.1 | AAO15382.1 | |||
| AJ272197.1 | CAB77268.1 | |||
| AK057202.1 | ||||
| AK126564.1 | ||||
| AK225448.1 | ||||
| BC015454.1 | AAH15454.1 | |||
| BC035325.1 | AAH35325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062194 | Chr.17:31755256 - 32041289 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map