Assay Details
Target Gene Details
Entrez Gene ID: | 192666 |
Gene Name: | keratin 24 |
Gene Aliases: |
K24, KA24 |
Location: |
Chr.17:40694246-40703750 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT24 | XM_017024299.1 | 7 | 1655 | XP_016879788.1 |
More Information
Set Membership: |
|

Genomic Map