Assay Details
Target Gene Details
| Entrez Gene ID: | 9487 | 
| Gene Name: | phosphatidylinositol glycan anchor biosynthesis class L | 
| Gene Aliases: | CHIME | 
| Location: | Chr.17:16217195-16351800 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| PIGL | NM_004278.3 | NP_004269.1 | ||
| XM_011524080.2 | XP_011522382.1 | |||
| XM_017025349.1 | XP_016880838.1 | |||
| XM_017025350.1 | XP_016880839.1 | |||
| XM_017025351.1 | XP_016880840.1 | |||
| XM_017025352.1 | XP_016880841.1 | |||
| XM_017025353.1 | XP_016880842.1 | |||
| XM_017025354.1 | XP_016880843.1 | |||
| XM_017025355.1 | XP_016880844.1 | |||
| XM_017025356.1 | XP_016880845.1 | |||
| AB017165.1 | BAA74775.1 | |||
| AK292932.1 | ||||
| AK296120.1 | ||||
| AK302523.1 | ||||
| BC068197.1 | AAH68197.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv528263 | Chr.17:16194116 - 16293682 on Build GRCh38 | Loss |  NCOR1  PIGL  MIR1288 | 
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |  ZSWIM7  CENPV  UBB  LOC101928567  ZNF624  TRPV2  TTC19  LRRC75A  ADORA2B  CCDC144A  ZNF287  SNORD65  SNORD49A  LRRC75A-AS1  CDRT15P2  NCOR1  SNORD49B  PIGL  MIR1288  MEIS3P1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top