Assay Details
Target Gene Details
Entrez Gene ID: | 9058 |
Gene Name: | solute carrier family 13 member 2 |
Gene Aliases: |
NADC1, NaCT, NaDC-1, SDCT1 |
Location: |
Chr.17:28473358-28497781 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC13A2 | NM_001145975.1 | NP_001139447.1 | ||
| NM_003984.3 | NP_003975.1 | |||
| NR_027384.1 | ||||
| XM_006722165.2 | XP_006722228.1 | |||
| XM_011525450.2 | XP_011523752.1 | |||
| XM_011525451.1 | XP_011523753.1 | |||
| XM_011525452.1 | XP_011523754.1 | |||
| XM_011525453.2 | XP_011523755.1 | |||
| XM_011525454.2 | XP_011523756.1 | |||
| AK091926.1 | ||||
| AK298388.1 | ||||
| AK303909.1 | ||||
| AK303936.1 | ||||
| AK314684.1 | ||||
| BC096276.1 | AAH96276.1 | |||
| BC096277.1 | AAH96277.1 | |||
| BC096278.1 | AAH96278.1 | |||
| BC096279.1 | AAH96279.1 | |||
| DB204108.1 | ||||
| U26209.1 | AAA98504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|
| nsv574653 | Chr.17:28227749 - 28501479 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map