Assay Details
Target Gene Details
Entrez Gene ID: | 55803 |
Gene Name: | ArfGAP with dual PH domains 2 |
Gene Aliases: |
CENTA2, HSA272195, cent-b |
Location: |
Chr.17:30921736-30959193 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAP2 | NM_018404.2 | NP_060874.1 | ||
| AJ238994.1 | CAB88383.1 | |||
| AJ242782.1 | CAC40651.1 | |||
| AJ272195.1 | CAB77266.1 | |||
| AK074196.1 | ||||
| AK074204.1 | ||||
| AK222526.1 | BAD96246.1 | |||
| BC033758.1 | AAH33758.1 | |||
| DQ288859.1 | ABB90599.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833412 | Chr.17:30793684 - 30984939 on Build GRCh38 | Loss |
|
| nsv457720 | Chr.17:30625268 - 31022670 on Build GRCh38 | Loss |
|
| esv2715813 | Chr.17:30601472 - 31009462 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map