Assay Details
Target Gene Details
Entrez Gene ID: | 6416 |
Gene Name: | mitogen-activated protein kinase kinase 4 |
Gene Aliases: |
JNKK, JNKK1, MAPKK4, MEK4, MKK4, PRKMK4, SAPKK-1, SAPKK1, SEK1, SERK1, SKK1 |
Location: |
Chr.17:12020818-12143831 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP2K4 | NM_001281435.1 | NP_001268364.1 | ||
| NM_003010.3 | NP_003001.1 | |||
| XM_005256755.2 | XP_005256812.1 | |||
| XM_011523976.2 | XP_011522278.1 | |||
| XM_011523977.2 | XP_011522279.1 | |||
| AK131544.1 | ||||
| AK294923.1 | ||||
| AK313053.1 | ||||
| BC036032.1 | AAH36032.1 | |||
| BC060764.1 | AAH60764.1 | |||
| BT019676.1 | AAV38482.1 | |||
| CR536564.1 | CAG38801.1 | |||
| L36870.1 | AAC41719.1 | |||
| U17743.1 | AAC50127.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25918 | Chr.17:12020685 - 12021704 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map