Assay Details
Target Gene Details
Entrez Gene ID: | 388403 |
Gene Name: | yippee like 2 |
Gene Aliases: |
FKSG4 |
Location: |
Chr.17:59331692-59401734 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YPEL2 | NM_001005404.3 | NP_001005404.1 | ||
| XM_017024621.1 | XP_016880110.1 | |||
| AB098736.1 | BAD51377.1 | |||
| AF305195.1 | AAL09354.1 | |||
| AI540782.1 | ||||
| BC127924.1 | ||||
| BC132884.1 | ||||
| BC132886.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833500 | Chr.17:59211557 - 59444989 on Build GRCh38 | Loss |
|
| nsv1062736 | Chr.17:59192781 - 59393940 on Build GRCh38 | Gain |
|
| nsv1057152 | Chr.17:59158105 - 59549885 on Build GRCh38 | Gain |
|
| esv3573764 | Chr.17:59142030 - 59424243 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map