Assay Details
Target Gene Details
Entrez Gene ID: | 7566 |
Gene Name: | zinc finger protein 18 |
Gene Aliases: |
HDSG1, KOX11, ZKSCAN6, ZNF535, ZSCAN38, Zfp535 |
Location: |
Chr.17:11957433-11997510 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF18 | NM_001303281.1 | NP_001290210.1 | ||
| NM_001303282.1 | NP_001290211.1 | |||
| NM_144680.3 | NP_653281.2 | |||
| XM_011524002.2 | XP_011522304.1 | |||
| XM_017025006.1 | XP_016880495.1 | |||
| XM_017025007.1 | XP_016880496.1 | |||
| XM_017025008.1 | XP_016880497.1 | |||
| XM_017025009.1 | XP_016880498.1 | |||
| XM_017025010.1 | XP_016880499.1 | |||
| XM_017025011.1 | XP_016880500.1 | |||
| AK057783.1 | BAB71570.1 | |||
| AK092656.1 | BAC03939.1 | |||
| AK127908.1 | ||||
| AY491053.1 | AAS67595.1 | |||
| BC036096.2 | AAH36096.1 |
More Information
Set Membership: |
|

Genomic Map