Assay Details
Target Gene Details
Entrez Gene ID: | 100289255 |
Gene Name: | long intergenic non-protein coding RNA 675 |
Gene Aliases: |
- |
Location: |
Chr.17:10794913-10804099 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00675 | NR_036581.1 | 2 | 728 | |
| BM924455.1 | 2 | 681 |
Target Gene Details
Entrez Gene ID: | 101101775 |
Gene Name: | TMEM220 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:10729777-10815164 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM220-AS1 | NR_073458.1 | |||
| DA190285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574351 | Chr.17:10762033 - 10814772 on Build GRCh38 | Loss |
|
| nsv833367 | Chr.17:10683841 - 10826633 on Build GRCh38 | Loss |
|
| nsv1058835 | Chr.17:10322963 - 10939602 on Build GRCh38 | Loss |
|

Genomic Map