Assay Details
Target Gene Details
Entrez Gene ID: | 2886 |
Gene Name: | growth factor receptor bound protein 7 |
Gene Aliases: |
- |
Location: |
Chr.17:39737909-39747285 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRB7 | NM_001030002.2 | NP_001025173.1 | ||
| NM_001242442.1 | NP_001229371.1 | |||
| NM_005310.3 | NP_005301.2 | |||
| XM_017024536.1 | 1 | 671 | XP_016880025.1 | |
| XM_017024537.1 | XP_016880026.1 | |||
| XM_017024538.1 | XP_016880027.1 | |||
| AK027729.1 | ||||
| AK125544.1 | ||||
| AK222849.1 | BAD96569.1 | |||
| AK222870.1 | BAD96590.1 | |||
| AK294620.1 | ||||
| AK314368.1 | ||||
| AU124875.1 | ||||
| BC006535.2 | AAH06535.1 | |||
| BE247550.1 | ||||
| D43772.1 | BAA07827.1 | |||
| DC384178.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953895 | Chr.17:39730748 - 39746847 on Build GRCh38 | Deletion |
|
| nsv1056234 | Chr.17:39706352 - 39753735 on Build GRCh38 | Gain |
|
| nsv2048 | Chr.17:39718682 - 39742588 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map