Assay Details
Target Gene Details
Entrez Gene ID: | 146664 |
Gene Name: | mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isozyme B |
Gene Aliases: |
GnT-IX, GnT-VB |
Location: |
Chr.17:76868487-76950389 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGAT5B | NM_001199172.1 | NP_001186101.1 | ||
| NM_144677.2 | NP_653278.2 | |||
| NM_198955.1 | NP_945193.1 | |||
| XM_006721707.3 | XP_006721770.1 | |||
| XM_011524350.2 | XP_011522652.1 | |||
| XM_011524352.2 | XP_011522654.1 | |||
| XM_011524354.2 | XP_011522656.1 | |||
| XM_017024200.1 | XP_016879689.1 | |||
| AB109185.1 | BAC84969.1 | |||
| AB114297.1 | BAD02406.1 | |||
| AB235153.1 | ||||
| BC062354.1 | AAH62354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457912 | Chr.17:76838238 - 76880546 on Build GRCh38 | Loss |
|
| nsv833550 | Chr.17:76712038 - 76886290 on Build GRCh38 | Loss |
|
| nsv1059991 | Chr.17:76802292 - 76896772 on Build GRCh38 | Loss |
|
| nsv576069 | Chr.17:76856392 - 76889866 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map