Assay Details
Target Gene Details
Entrez Gene ID: | 51393 |
Gene Name: | transient receptor potential cation channel subfamily V member 2 |
Gene Aliases: |
VRL, VRL-1, VRL1 |
Location: |
Chr.17:16415542-16437003 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPV2 | NM_016113.4 | NP_057197.2 | ||
| XM_005256676.1 | XP_005256733.1 | |||
| XM_005256677.1 | XP_005256734.1 | |||
| XM_005256678.4 | XP_005256735.1 | |||
| XM_006721541.3 | XP_006721604.1 | |||
| XM_006721543.3 | XP_006721606.1 | |||
| XM_011523922.1 | XP_011522224.1 | |||
| XM_011523923.1 | XP_011522225.1 | |||
| XM_017024730.1 | XP_016880219.1 | |||
| XM_017024731.1 | XP_016880220.1 | |||
| XM_017024732.1 | XP_016880221.1 | |||
| AF103906.1 | AAD41724.1 | |||
| AF129112.1 | AAD26363.1 | |||
| AJ487963.1 | CAD32310.1 | |||
| AK001896.1 | ||||
| AK289705.1 | ||||
| BC018926.2 | AAH18926.1 | |||
| BC051305.1 | AAH51305.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063764 | Chr.17:16268335 - 16759600 on Build GRCh38 | Loss |
|
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map