Assay Details
Target Gene Details
Entrez Gene ID: | 11322 |
Gene Name: | transmembrane channel like 6 |
Gene Aliases: |
EV1, EVER1, EVIN1, LAK-4P |
Location: |
Chr.17:78108775-78132427 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMC6 | AK074065.1 | 20 | 4201 |
Target Gene Details
Entrez Gene ID: | 100131096 |
Gene Name: | TNRC6C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:78107401-78111799 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNRC6C-AS1 | NR_040071.1 | 1 | 275 | |
| AK123771.1 | 1 | 453 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833554 | Chr.17:77977426 - 78151914 on Build GRCh38 | Gain+Loss |
|

Genomic Map