Assay Details
Target Gene Details
Entrez Gene ID: | 162427 |
Gene Name: | family with sequence similarity 134 member C |
Gene Aliases: |
- |
Location: |
Chr.17:42579508-42609427 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM134C | NM_178126.3 | NP_835227.1 | ||
| NR_026697.1 | ||||
| XM_011524440.2 | XP_011522742.1 | |||
| AK091125.1 | ||||
| AK225797.1 | ||||
| AK294791.1 | ||||
| AK308288.1 | ||||
| BC049370.1 | AAH49370.1 | |||
| BX647175.1 | ||||
| DB289370.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map