Assay Details
Target Gene Details
Entrez Gene ID: | 116236 |
Gene Name: | abhydrolase domain containing 15 |
Gene Aliases: |
- |
Location: |
Chr.17:29560661-29567024 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD15 | NM_198147.2 | NP_937790.2 | ||
| AK056717.1 | ||||
| AY358212.1 | AAQ88579.1 | |||
| BC012476.1 | AAH12476.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521557 | Chr.17:29562968 - 29595257 on Build GRCh38 | Loss |
|
| nsv833409 | Chr.17:29465646 - 29655807 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map