Assay Details
Target Gene Details
Entrez Gene ID: | 388815 |
Gene Name: | mir-99a-let-7c cluster host gene |
Gene Aliases: |
C21orf34, C21orf35, LINC00478, MONC |
Location: |
Chr.21:16070488-16631727 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR99AHG | NR_027790.3 | |||
| NR_111004.2 | ||||
| NR_111005.2 | ||||
| NR_136541.1 | ||||
| NR_136542.1 | ||||
| AF486622.1 | ||||
| AK095614.1 | ||||
| CN313455.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74959454] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map